Anti-WISP1 Mouse Monoclonal Antibody [clone: 213611]

Supplier: R & D Systems
MAB1627
220019-222EA 772.25 USD
220019-222
Anti-WISP1 Mouse Monoclonal Antibody [clone: 213611]
Antibodies
Type: Primary
Antigen: WISP1
Clonality: Monoclonal
Clone: 213611
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR