Anti-Tenascin R Mouse Monoclonal Antibody [clone: 619]

Supplier: R & D Systems
MAB1624
220019-221EA 600.15 USD
220019-221
Anti-Tenascin R Mouse Monoclonal Antibody [clone: 619]
Antibodies
Type: Primary
Antigen: Tenascin R
Clonality: Monoclonal
Clone: 619
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR