Anti-CD209 Mouse Monoclonal Antibody [clone: DC28]

Supplier: R & D Systems
MAB16211
220019-220EA 772.25 USD
220019-220
Anti-CD209 Mouse Monoclonal Antibody [clone: DC28]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: DC28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR