Anti-CLEC4M Mouse Monoclonal Antibody [clone: 120604]

Supplier: R & D Systems
MAB162
220019-218EA 763.57 USD
220019-218
Anti-CLEC4M Mouse Monoclonal Antibody [clone: 120604]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: CLEC4M
Clonality: Monoclonal
Clone: 120604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR