Anti-SEMA6A Rat Monoclonal Antibody [clone: 201932]

Supplier: R & D Systems
MAB1615
220019-216EA 763.57 USD
220019-216
Anti-SEMA6A Rat Monoclonal Antibody [clone: 201932]
Antibodies
Type: Primary
Antigen: SEMA6A
Clonality: Monoclonal
Clone: 201932
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR