Anti-FGL1 Mouse Monoclonal Antibody [clone: 198401]

Supplier: R & D Systems
MAB1614
220019-215EA 772.25 USD
220019-215
Anti-FGL1 Mouse Monoclonal Antibody [clone: 198401]
Antibodies
Type: Primary
Antigen Symbol: FGL1
Antigen Name: fibrinogen-like 1
Antigen Synonyms: HFREP1, HP-041, LFIRE-1, LFIRE1
Clonality: Monoclonal
Clone: 198401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR