Anti-TNFRSF12A Rat Monoclonal Antibody [clone: 314615]

Supplier: R & D Systems
MAB1610
220019-213EA 594.37 USD
220019-213
Anti-TNFRSF12A Rat Monoclonal Antibody [clone: 314615]
Antibodies
Type: Primary
Antigen: TNFRSF12A
Clonality: Monoclonal
Clone: 314615
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR