Anti-IL6 Mouse Monoclonal Antibody [clone: 247002]

Supplier: R & D Systems
MAB16091
220019-211EA 763.57 USD
220019-211
Anti-IL6 Mouse Monoclonal Antibody [clone: 247002]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 247002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR