Anti-IL6 Mouse Monoclonal Antibody [clone: 247017]

Supplier: R & D Systems
MAB1609
220019-210EA 763.57 USD
220019-210
Anti-IL6 Mouse Monoclonal Antibody [clone: 247017]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 247017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR