Anti-CXCL8 Mouse Monoclonal Antibody [clone: 258901]

Supplier: R & D Systems
MAB16081
220019-209EA 772.25 USD
220019-209
Anti-CXCL8 Mouse Monoclonal Antibody [clone: 258901]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 258901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR