Anti-CXCL8 Mouse Monoclonal Antibody [clone: 258911]

Supplier: R & D Systems
MAB1608
220019-208EA 594.37 USD
220019-208
Anti-CXCL8 Mouse Monoclonal Antibody [clone: 258911]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 258911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR