Anti-CXCR3 Mouse Monoclonal Antibody [clone: 49801]

Supplier: R & D Systems
MAB160-100
102885-278EA 655.11 USD
102885-278
Anti-CXCR3 Mouse Monoclonal Antibody [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR