Anti-KLK11 Rat Monoclonal Antibody [clone: 371604]

Supplier: R & D Systems
MAB15951
220019-199EA 577.02 USD
220019-199
Anti-KLK11 Rat Monoclonal Antibody [clone: 371604]
Antibodies
Type: Primary
Antigen: KLK11
Clonality: Monoclonal
Clone: 371604
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR