Anti-FGFBP1 Mouse Monoclonal Antibody [clone: 196519]

Supplier: R & D Systems
MAB1593
220019-197EA 763.57 USD
220019-197
Anti-FGFBP1 Mouse Monoclonal Antibody [clone: 196519]
Antibodies
Type: Primary
Antigen Symbol: FGFBP1
Antigen Name: fibroblast growth factor binding protein 1
Antigen Synonyms: FGF-BP, FGF-BP1, FGFBP, FGFBP-1, HBP17
Clonality: Monoclonal
Clone: 196519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR