Anti-DPP Mouse Monoclonal Antibody [clone: 146609]

Supplier: R & D Systems
MAB159
220019-196EA 763.57 USD
220019-196
Anti-DPP Mouse Monoclonal Antibody [clone: 146609]
Antibodies
Type: Primary
Antigen: DPP
Clonality: Monoclonal
Clone: 146609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR