Anti-SOST Rat Monoclonal Antibody [clone: 248121]

Supplier: R & D Systems
MAB1589
220019-195EA 763.57 USD
220019-195
Anti-SOST Rat Monoclonal Antibody [clone: 248121]
Antibodies
Type: Primary
Antigen: SOST
Clonality: Monoclonal
Clone: 248121
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR