Anti-IFNL2 Mouse Monoclonal Antibody [clone: 248526]

Supplier: R & D Systems
MAB1587
220019-193EA 647.88 USD
220019-193
Anti-IFNL2 Mouse Monoclonal Antibody [clone: 248526]
Antibodies
Type: Primary
Antigen: IFNL2
Clonality: Monoclonal
Clone: 248526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR