Anti-TMPRSS15 Mouse Monoclonal Antibody [clone: 190034]

Supplier: R & D Systems
MAB1585
220019-192EA 763.57 USD
220019-192
Anti-TMPRSS15 Mouse Monoclonal Antibody [clone: 190034]
Antibodies
Type: Primary
Antigen: TMPRSS15
Clonality: Monoclonal
Clone: 190034
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR