Anti-STAT5B Mouse Monoclonal Antibody [clone: 389215]

Supplier: R & D Systems
MAB1584
220019-191EA 594.37 USD
220019-191
Anti-STAT5B Mouse Monoclonal Antibody [clone: 389215]
Antibodies
Type: Primary
Antigen: STAT5B
Clonality: Monoclonal
Clone: 389215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR