Anti-HGFAC Mouse Monoclonal Antibody [clone: 196817]

Supplier: R & D Systems
MAB1583
220019-190EA 763.57 USD
220019-190
Anti-HGFAC Mouse Monoclonal Antibody [clone: 196817]
Antibodies
Type: Primary
Antigen: HGFAC
Clonality: Monoclonal
Clone: 196817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR