Anti-ICOSL Rat Monoclonal Antibody [clone: 599841]

Supplier: R & D Systems
MAB158
102885-276EA 594.37 USD
102885-276
Anti-ICOSL Rat Monoclonal Antibody [clone: 599841]
Antibodies
Type: Primary
Antigen: ICOSL
Clonality: Monoclonal
Clone: 599841
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR