Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 631122]

Supplier: R & D Systems
MAB15761
102885-274EA 594.37 USD
102885-274
Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 631122]
Antibodies
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Monoclonal
Clone: 631122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR