Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 216703]

Supplier: R & D Systems
MAB1576
220019-189EA 639.2 USD
220019-189
Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 216703]
Antibodies
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Monoclonal
Clone: 216703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR