Anti-NBN Mouse Monoclonal Antibody [clone: 206911]

Supplier: R & D Systems
MAB15731
220019-188EA 577.02 USD
220019-188
Anti-NBN Mouse Monoclonal Antibody [clone: 206911]
Antibodies
Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 206911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR