Anti-NBN Mouse Monoclonal Antibody [clone: 206919]

Supplier: R & D Systems
MAB1573
220019-187EA 566.89 USD
220019-187
Anti-NBN Mouse Monoclonal Antibody [clone: 206919]
Antibodies
Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 206919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR