Anti-KNG1 Mouse Monoclonal Antibody [clone: 236012]

Supplier: R & D Systems
MAB15692
220019-185EA 772.25 USD
220019-185
Anti-KNG1 Mouse Monoclonal Antibody [clone: 236012]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 236012
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR