Anti-KNG1 Mouse Monoclonal Antibody [clone: 236006]

Supplier: R & D Systems
MAB15691
220019-184EA 772.25 USD
220019-184
Anti-KNG1 Mouse Monoclonal Antibody [clone: 236006]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 236006
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR