Anti-KNG1 Mouse Monoclonal Antibody [clone: 207025]

Supplier: R & D Systems
MAB1569
220019-183EA 763.57 USD
220019-183
Anti-KNG1 Mouse Monoclonal Antibody [clone: 207025]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 207025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR