Anti-HHIP Rat Monoclonal Antibody [clone: 217413]

Supplier: R & D Systems
MAB1568
220019-182EA 763.57 USD
220019-182
Anti-HHIP Rat Monoclonal Antibody [clone: 217413]
Antibodies
Type: Primary
Antigen: HHIP
Clonality: Monoclonal
Clone: 217413
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR