Anti-GH1 Mouse Monoclonal Antibody [clone: 222517]

Supplier: R & D Systems
MAB15661
220019-180EA 763.57 USD
220019-180
Anti-GH1 Mouse Monoclonal Antibody [clone: 222517]
Antibodies
Type: Primary
Antigen Symbol: GH1
Antigen Name: Growth Hormone
Antigen Synonyms: Somatotropin
Clonality: Monoclonal
Clone: 222517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR