Anti-GH1 Mouse Monoclonal Antibody [clone: 222540]

Supplier: R & D Systems
MAB1566
220019-179EA 772.25 USD
220019-179
Anti-GH1 Mouse Monoclonal Antibody [clone: 222540]
Antibodies
Type: Primary
Antigen Symbol: GH1
Antigen Name: Growth Hormone
Antigen Synonyms: Somatotropin
Clonality: Monoclonal
Clone: 222540
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR