Anti-FABP1 Mouse Monoclonal Antibody [clone: 220119]

Supplier: R & D Systems
MAB1565
220019-178EA 561.11 USD
220019-178
Anti-FABP1 Mouse Monoclonal Antibody [clone: 220119]
Antibodies
Type: Primary
Antigen Symbol: FABP1
Antigen Name: fatty acid binding protein 1, liver
Antigen Synonyms: Fabplg, L-FABP, SCP, SCP., p14
Clonality: Monoclonal
Clone: 220119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR