Anti-OLR1 Rat Monoclonal Antibody [clone: 214001]

Supplier: R & D Systems
MAB15641
220019-177EA 763.57 USD
220019-177
Anti-OLR1 Rat Monoclonal Antibody [clone: 214001]
Antibodies
Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: 214001
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR