Anti-AHSG Rat Monoclonal Antibody [clone: 2744]

Supplier: R & D Systems
MAB15632
220019-175EA 772.25 USD
220019-175
Anti-AHSG Rat Monoclonal Antibody [clone: 2744]
Antibodies
Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 2744
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR