Anti-AHSG Rat Monoclonal Antibody [clone: 27431]

Supplier: R & D Systems
MAB1563
220019-174EA 763.57 USD
220019-174
Anti-AHSG Rat Monoclonal Antibody [clone: 27431]
Antibodies
Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 27431
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR