Anti-CD274 Mouse Monoclonal Antibody [clone: 130021]

Supplier: R & D Systems
MAB1561
220019-173EA 655.11 USD
220019-173
Anti-CD274 Mouse Monoclonal Antibody [clone: 130021]
Antibodies
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 130021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR