Anti-CCR3 Rat Monoclonal Antibody [clone: 83103]

Supplier: R & D Systems
MAB1551-100
102885-140EA 633.42 USD
102885-140
Anti-CCR3 Rat Monoclonal Antibody [clone: 83103]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 83103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR