Anti-CCR3 Rat Monoclonal Antibody [clone: 61828]

Supplier: R & D Systems
MAB155-100
102885-138EA 639.2 USD
102885-138
Anti-CCR3 Rat Monoclonal Antibody [clone: 61828]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 61828
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR