Anti-DKKL1 Mouse Monoclonal Antibody [clone: 202509]

Supplier: R & D Systems
MAB1549
220019-168EA 772.25 USD
220019-168
Anti-DKKL1 Mouse Monoclonal Antibody [clone: 202509]
Antibodies
Type: Primary
Antigen: DKKL1
Clonality: Monoclonal
Clone: 202509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR