Anti-CSF2 Mouse Monoclonal Antibody [clone: 249201]

Supplier: R & D Systems
MAB1546
220019-166EA 763.57 USD
220019-166
Anti-CSF2 Mouse Monoclonal Antibody [clone: 249201]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 249201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR