Anti-TNFRSF10B Rat Monoclonal Antibody [clone: 118929]

Supplier: R & D Systems
MAB1540
220019-163EA 763.57 USD
220019-163
Anti-TNFRSF10B Rat Monoclonal Antibody [clone: 118929]
Antibodies
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 118929
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR