Anti-TDGF1 Rat Monoclonal Antibody [clone: 237614]

Supplier: R & D Systems
MAB15381
220019-161EA 763.57 USD
220019-161
Anti-TDGF1 Rat Monoclonal Antibody [clone: 237614]
Antibodies
Type: Primary
Antigen: TDGF1
Clonality: Monoclonal
Clone: 237614
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR