Anti-TDGF1 Rat Monoclonal Antibody [clone: 237603]

Supplier: R & D Systems
MAB1538
220019-160EA 527.85 USD
220019-160
Anti-TDGF1 Rat Monoclonal Antibody [clone: 237603]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TDGF1
Clonality: Monoclonal
Clone: 237603
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR