Anti-MTOR Rat Monoclonal Antibody [clone: 303728]

Supplier: R & D Systems
MAB1537
220019-159EA 594.37 USD
220019-159
Anti-MTOR Rat Monoclonal Antibody [clone: 303728]
Antibodies
Type: Primary
Antigen: MTOR
Clonality: Monoclonal
Clone: 303728
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR