Anti-HIF1A Mouse Monoclonal Antibody [clone: 241809]

Supplier: R & D Systems
MAB1536
220019-158EA 647.88 USD
220019-158
Anti-HIF1A Mouse Monoclonal Antibody [clone: 241809]
Antibodies
Simple Western

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: 241809
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR