Anti-SELL Mouse Monoclonal Antibody [clone: 196021]

Supplier: R & D Systems
MAB1534
220019-157EA 763.57 USD
220019-157
Anti-SELL Mouse Monoclonal Antibody [clone: 196021]
Antibodies
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 196021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR