Anti-TLR6 Rat Monoclonal Antibody [clone: 418601]

Supplier: R & D Systems
MAB1533
220019-156EA 577.02 USD
220019-156
Anti-TLR6 Rat Monoclonal Antibody [clone: 418601]
Antibodies
Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone: 418601
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR