Anti-CD209E Rat Monoclonal Antibody [clone: 210006]

Supplier: R & D Systems
MAB1528
220019-153EA 763.57 USD
220019-153
Anti-CD209E Rat Monoclonal Antibody [clone: 210006]
Antibodies
Type: Primary
Antigen: CD209E
Clonality: Monoclonal
Clone: 210006
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR