Anti-PRL7D1 Rat Monoclonal Antibody [clone: 219310]

Supplier: R & D Systems
MAB1527
220019-152EA 763.57 USD
220019-152
Anti-PRL7D1 Rat Monoclonal Antibody [clone: 219310]
Antibodies
Type: Primary
Antigen: PRL7D1
Clonality: Monoclonal
Clone: 219310
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR