Anti-FZD6 Rat Monoclonal Antibody [clone: 211435]

Supplier: R & D Systems
MAB1526-050 MAB1526-500
220019-151EA 763.57 USD
220019-151 220019-150
Anti-FZD6 Rat Monoclonal Antibody [clone: 211435]
Antibodies
Type: Primary
Antigen Symbol: FZD6
Antigen Name: frizzled homolog 6 (Drosophila)
Antigen Synonyms: Fz6
Clonality: Monoclonal
Clone: 211435
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR